missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Dioxene 98.0+%, TCI America™
$333.46 - $2899.94
Chemical Identifiers
CAS | 543-75-9 |
---|---|
Molecular Formula | C4H6O2 |
Molecular Weight (g/mol) | 86.09 |
MDL Number | MFCD00082537 |
InChI Key | HIZVCIIORGCREW-UHFFFAOYSA-N |
Synonym | 1,4-dioxene, 1,4-dioxin, 2,3-dihydro, p-dioxin, 2,3-dihydro, 2,3-dihydro-1,4-dioxin, 1,4-dioxane-2,3-diyl, # |
PubChem CID | 136353 |
IUPAC Name | 2,3-dihydro-1,4-dioxine |
SMILES | C1COC=CO1 |
Chemical Identifiers
543-75-9 | |
86.09 | |
HIZVCIIORGCREW-UHFFFAOYSA-N | |
136353 | |
C1COC=CO1 |
C4H6O2 | |
MFCD00082537 | |
1,4-dioxene, 1,4-dioxin, 2,3-dihydro, p-dioxin, 2,3-dihydro, 2,3-dihydro-1,4-dioxin, 1,4-dioxane-2,3-diyl, # | |
2,3-dihydro-1,4-dioxine |
Specifications
543-75-9 | |
94°C | |
MFCD00082537 | |
1993 | |
HIZVCIIORGCREW-UHFFFAOYSA-N | |
2,3-dihydro-1,4-dioxine | |
136353 | |
≥98.0% (GC) | |
1,4-Dioxene |
Yellow | |
C4H6O2 | |
1 g | |
1,4-dioxene, 1,4-dioxin, 2,3-dihydro, p-dioxin, 2,3-dihydro, 2,3-dihydro-1,4-dioxin, 1,4-dioxane-2,3-diyl, # | |
C1COC=CO1 | |
86.09 | |
86.09 | |
Liquid |
Safety and Handling
TSCA : No