missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Hexadiene (cis- and trans- mixture) 99.0+%, TCI America™

Supplier:  TCI America H03335ML

 View more versions of this product

Catalog No. H03335ML


Explore available promotions

Description

Description

Specifications

Specifications

1,4-Hexadiene (cis- and trans- mixture)
Yellow
C6H10
5 mL
1,4-hexadiene, trans-1,4-hexadiene, 1,4-hexadiene, 4e, 1,4-hexadiene,c&t, allylpropenyl, 1,4-hexadiene cis-trans, 1,4-hexa-1,4-diene, 1-allylpropene, 1,4-hexadiene,cis-trans, 1,4-trans-hexadiene
CC=CCC=C
82.146
82.15
Liquid
592-45-0
65°C
MFCD00009298
2458
PRBHEGAFLDMLAL-GQCTYLIASA-N
(4E)-hexa-1,4-diene
5365552
≥99.0% (GC)

Chemical Identifiers

592-45-0
82.146
PRBHEGAFLDMLAL-GQCTYLIASA-N
5365552
CC=CCC=C
C6H10
MFCD00009298
1,4-hexadiene, trans-1,4-hexadiene, 1,4-hexadiene, 4e, 1,4-hexadiene,c&t, allylpropenyl, 1,4-hexadiene cis-trans, 1,4-hexa-1,4-diene, 1-allylpropene, 1,4-hexadiene,cis-trans, 1,4-trans-hexadiene
(4E)-hexa-1,4-diene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions