missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Methoxyphenyl)-5-methyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 187998-64-7
Molecular Formula C12H12N2O3
Molecular Weight (g/mol) 232.239
MDL Number MFCD02677745
InChI Key JPFGKGZYCXLEGQ-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl-5-methyl-1h-pyrazole-4-carboxylic acid, 1-4-methoxyphenyl-5-methylpyrazole-4-carboxylic acid, 1-4-methoxyphenyl-5-methyl-1h-pyrazole-4-carboxylicacid, 1h-pyrazole-4-carboxylicacid, 1-4-methoxyphenyl-5-methyl, 1h-pyrazole-4-carboxylicacid,1-4-methoxyphenyl-5-methyl
PubChem CID 2776133
IUPAC Name 1-(4-methoxyphenyl)-5-methylpyrazole-4-carboxylic acid
SMILES CC1=C(C=NN1C2=CC=C(C=C2)OC)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC00105CB
SDS View Documents
Thermo Scientific Chemicals
CC00105CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC00105DA
SDS View Documents
Thermo Scientific Chemicals
CC00105DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

187998-64-7
232.239
JPFGKGZYCXLEGQ-UHFFFAOYSA-N
2776133
CC1=C(C=NN1C2=CC=C(C=C2)OC)C(=O)O
C12H12N2O3
MFCD02677745
1-4-methoxyphenyl-5-methyl-1h-pyrazole-4-carboxylic acid, 1-4-methoxyphenyl-5-methylpyrazole-4-carboxylic acid, 1-4-methoxyphenyl-5-methyl-1h-pyrazole-4-carboxylicacid, 1h-pyrazole-4-carboxylicacid, 1-4-methoxyphenyl-5-methyl, 1h-pyrazole-4-carboxylicacid,1-4-methoxyphenyl-5-methyl
1-(4-methoxyphenyl)-5-methylpyrazole-4-carboxylic acid

Specifications

187998-64-7
100.0
Amber Glass Bottle
MFCD02677745
1-4-methoxyphenyl-5-methyl-1h-pyrazole-4-carboxylic acid, 1-4-methoxyphenyl-5-methylpyrazole-4-carboxylic acid, 1-4-methoxyphenyl-5-methyl-1h-pyrazole-4-carboxylicacid, 1h-pyrazole-4-carboxylicacid, 1-4-methoxyphenyl-5-methyl, 1h-pyrazole-4-carboxylicacid,1-4-methoxyphenyl-5-methyl
CC1=C(C=NN1C2=CC=C(C=C2)OC)C(=O)O
232.239
232.24
1-(4-Methoxyphenyl)-5-methyl-1H-pyrazole-4-carboxylic acid
96.5
97%
C12H12N2O3
250mg
JPFGKGZYCXLEGQ-UHFFFAOYSA-N
1-(4-methoxyphenyl)-5-methylpyrazole-4-carboxylic acid
2776133
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'