missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Methylbenzoyl)pyrrolidine, 97%, Thermo Scientific™

$741.42 - $741.42

Chemical Identifiers

CAS 59746-40-6
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD00996306
InChI Key QXMOFZGGGOFRPH-UHFFFAOYSA-N
Synonym 1-4-methylbenzoyl pyrrolidine, 4-methylphenyl 1-pyrrolidinyl methanone, aronis25719, 4-pyrrolidinecarbonyl toluene, 4-methylphenyl pyrrolidinyl ketone, 1-4-methylphenyl carbonyl pyrrolidine, 4-methylphenyl-pyrrolidin-1-ylmethanone, 4-methylphenyl pyrrolidin-1-yl methanone
PubChem CID 721203
IUPAC Name (4-methylphenyl)-pyrrolidin-1-ylmethanone
SMILES CC1=CC=C(C=C1)C(=O)N2CCCC2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5603703
View Documents
Thermo Scientific Chemicals
H5603703
1 g
Each for $741.42
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

59746-40-6
189.258
QXMOFZGGGOFRPH-UHFFFAOYSA-N
721203
CC1=CC=C(C=C1)C(=O)N2CCCC2
C12H15NO
MFCD00996306
1-4-methylbenzoyl pyrrolidine, 4-methylphenyl 1-pyrrolidinyl methanone, aronis25719, 4-pyrrolidinecarbonyl toluene, 4-methylphenyl pyrrolidinyl ketone, 1-4-methylphenyl carbonyl pyrrolidine, 4-methylphenyl-pyrrolidin-1-ylmethanone, 4-methylphenyl pyrrolidin-1-yl methanone
(4-methylphenyl)-pyrrolidin-1-ylmethanone

Specifications

59746-40-6
MFCD00996306
1-4-methylbenzoyl pyrrolidine, 4-methylphenyl 1-pyrrolidinyl methanone, aronis25719, 4-pyrrolidinecarbonyl toluene, 4-methylphenyl pyrrolidinyl ketone, 1-4-methylphenyl carbonyl pyrrolidine, 4-methylphenyl-pyrrolidin-1-ylmethanone, 4-methylphenyl pyrrolidin-1-yl methanone
CC1=CC=C(C=C1)C(=O)N2CCCC2
189.258
189.25
1-(4-Methylbenzoyl)pyrrolidine
C12H15NO
1 g
QXMOFZGGGOFRPH-UHFFFAOYSA-N
(4-methylphenyl)-pyrrolidin-1-ylmethanone
721203
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'