missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Phenylenediamine 98.0+%, TCI America™

Supplier:  TCI America P017025G

 View more versions of this product

Catalog No. P017025G


Explore available promotions

Description

Description

Specifications

Specifications

1,4-Phenylenediamine
142°C
267°C
MFCD00007901
1673
CBCKQZAAMUWICA-UHFFFAOYSA-N
benzene-1,4-diamine
7814
108.14
Crystalline Powder
106-50-3
Red-White
C6H8N2
25 g
p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine
C1=CC(=CC=C1N)N
108.144
CHEBI:51403
≥98.0% (GC,T)

Chemical Identifiers

106-50-3
108.144
CBCKQZAAMUWICA-UHFFFAOYSA-N
7814
benzene-1,4-diamine
C6H8N2
MFCD00007901
p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine
CHEBI:51403
C1=CC(=CC=C1N)N
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0185&(5)-4998

RTECSNumber : SS8050000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions