Learn More
1-(4-Pyridyl)ethylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5030303
Description
1-(4-Pyridyl)ethylamine is used as a pharmaceutical intermediates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(4-Pyridyl)ethylamine is used as a pharmaceutical intermediates
Solubility
Soluble in water
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
1-(4-Pyridyl)ethylamine | |
C7H11N2 | |
1 g | |
Air Sensitive | |
Soluble in water | |
C[C@H]([NH3+])C1=CC=NC=C1 | |
123.18 | |
122.17 |
50392-78-4 | |
MFCD02245004,MFCD09256818,MFCD09256819 | |
UN2735 | |
1-4-pyridyl ethylamine, 1-pyridin-4-yl-ethylamine, 1-pyridin-4-yl ethanamine, 1-4-pyridinyl ethanamine, 1-pyridin-4-yl ethan-1-amine, 1-4-pyridinyl ethanamin, 4-1-aminoethyl pyridine, alpha-methyl-4-pyridinemethylamine, 1-pyridin-4-ylethylamine, acmc-1aq0e | |
HIZMJYQEHFJWQY-LURJTMIESA-O | |
1-pyridin-4-ylethanamine | |
247998 | |
97% |
Chemical Identifiers
50392-78-4 | |
123.18 | |
HIZMJYQEHFJWQY-LURJTMIESA-O | |
247998 | |
C[C@H]([NH3+])C1=CC=NC=C1 |
C7H11N2 | |
MFCD02245004,MFCD09256818,MFCD09256819 | |
1-4-pyridyl ethylamine, 1-pyridin-4-yl-ethylamine, 1-pyridin-4-yl ethanamine, 1-4-pyridinyl ethanamine, 1-pyridin-4-yl ethan-1-amine, 1-4-pyridinyl ethanamin, 4-1-aminoethyl pyridine, alpha-methyl-4-pyridinemethylamine, 1-pyridin-4-ylethylamine, acmc-1aq0e | |
1-pyridin-4-ylethanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only