missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Pyridyl)ethylamine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H50303MD

 View more versions of this product

Catalog No. AAH50303MD


Explore available promotions

Description

Description

1-(4-Pyridyl)ethylamine is used as a pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Pyridyl)ethylamine is used as a pharmaceutical intermediates

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

1-(4-Pyridyl)ethylamine
C7H11N2
250 mg
Air Sensitive
Soluble in water
C[C@H]([NH3+])C1=CC=NC=C1
123.18
122.17
50392-78-4
MFCD02245004,MFCD09256818,MFCD09256819
UN2735
1-4-pyridyl ethylamine, 1-pyridin-4-yl-ethylamine, 1-pyridin-4-yl ethanamine, 1-4-pyridinyl ethanamine, 1-pyridin-4-yl ethan-1-amine, 1-4-pyridinyl ethanamin, 4-1-aminoethyl pyridine, alpha-methyl-4-pyridinemethylamine, 1-pyridin-4-ylethylamine, acmc-1aq0e
HIZMJYQEHFJWQY-LURJTMIESA-O
1-pyridin-4-ylethanamine
247998
97%

Chemical Identifiers

50392-78-4
123.18
HIZMJYQEHFJWQY-LURJTMIESA-O
247998
C[C@H]([NH3+])C1=CC=NC=C1
C7H11N2
MFCD02245004,MFCD09256818,MFCD09256819
1-4-pyridyl ethylamine, 1-pyridin-4-yl-ethylamine, 1-pyridin-4-yl ethanamine, 1-4-pyridinyl ethanamine, 1-pyridin-4-yl ethan-1-amine, 1-4-pyridinyl ethanamin, 4-1-aminoethyl pyridine, alpha-methyl-4-pyridinemethylamine, 1-pyridin-4-ylethylamine, acmc-1aq0e
1-pyridin-4-ylethanamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only