Learn More
1,5-Bis(diphenylphosphino)pentane, 97%, Thermo Scientific Chemicals
$74.51 - $1166.93
Chemical Identifiers
CAS | 27721-02-4 |
---|---|
Molecular Formula | C29H30P2 |
Molecular Weight (g/mol) | 440.51 |
MDL Number | MFCD00003052 |
InChI Key | MZFPAWGWFDGCHP-UHFFFAOYSA-N |
Synonym | 1,5-bis diphenylphosphino pentane, dpppe, dppent, 5-diphenylphosphanylpentyl diphenyl phosphane, pentamethylenebis diphenylphosphine, 1,5-pentanediylbis diphenylphosphine, phosphine, 1,5-pentanediylbis diphenyl, 1-5-diphenylphosphino pentyl phenyl phosphino benzene |
PubChem CID | 2733414 |
SMILES | C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2105403
|
Thermo Scientific Chemicals
B2105403 |
1 g |
Each for $74.51
|
|
|||||
AAB2105406
|
Thermo Scientific Chemicals
B2105406 |
5 g |
Each for $290.36
|
|
|||||
AAB2105414
|
Thermo Scientific Chemicals
B2105414 |
25 g |
Each for $1,166.93
|
|
|||||
Description
1,5-Bis(diphenylphosphino)pentane is used as a ligand for the cross coupling reactions. It is used in the manufacturing of glass bottle PTFE bottle, aluminum foil bag and in cardboard drum. combination of a 1,5-bis(diphenylphosphino)pentane ligand and the addition of N,N,N?,N?-tetramethylethylenediamine (TMEDA) as a co-catalyst are the key factors in obtaining the corresponding aryl nitriles with improved catalyst productivities and selectivities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,5-Bis(diphenylphosphino)pentane is used as a ligand for the cross coupling reactions. It is used in the manufacturing of glass bottle PTFE bottle, aluminum foil bag and in cardboard drum. combination of a 1,5-bis(diphenylphosphino)pentane ligand and the addition of N,N,N′,N′-tetramethylethylenediamine (TMEDA) as a co-catalyst are the key factors in obtaining the corresponding aryl nitriles with improved catalyst productivities and selectivities.
Solubility
Soluble in N,N-dimethyl formamide.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
27721-02-4 | |
440.51 | |
MZFPAWGWFDGCHP-UHFFFAOYSA-N | |
2733414 |
C29H30P2 | |
MFCD00003052 | |
1,5-bis diphenylphosphino pentane, dpppe, dppent, 5-diphenylphosphanylpentyl diphenyl phosphane, pentamethylenebis diphenylphosphine, 1,5-pentanediylbis diphenylphosphine, phosphine, 1,5-pentanediylbis diphenyl, 1-5-diphenylphosphino pentyl phenyl phosphino benzene | |
C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
27721-02-4 | |
C29H30P2 | |
1 g | |
Soluble in N,N-dimethyl formamide. | |
C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1 | |
440.51 | |
440.5 | |
1,5-Bis(diphenylphosphino)pentane |
46°C to 48°C | |
MFCD00003052 | |
1,5-bis diphenylphosphino pentane, dpppe, dppent, 5-diphenylphosphanylpentyl diphenyl phosphane, pentamethylenebis diphenylphosphine, 1,5-pentanediylbis diphenylphosphine, phosphine, 1,5-pentanediylbis diphenyl, 1-5-diphenylphosphino pentyl phenyl phosphino benzene | |
MZFPAWGWFDGCHP-UHFFFAOYSA-N | |
5-diphenylphosphanylpentyl(diphenyl)phosphane | |
2733414 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only