Learn More
1,5-Hexadiene, 98%, Thermo Scientific Chemicals
$68.86 - $560.85
Chemical Identifiers
CAS | 592-42-7 |
---|---|
Molecular Formula | C6H10 |
Molecular Weight (g/mol) | 82.146 |
MDL Number | MFCD00008666 |
InChI Key | PYGSKMBEVAICCR-UHFFFAOYSA-N |
Synonym | 1,5-hexadiene, biallyl, diallyl, hexadiene dot, alpha,omega-hexadiene, .alpha.,.omega.-hexadiene, unii-4mtz4764fi, 1,5-hexanediene, acmc-1axer, hexa-1,5-dien-1-yl |
PubChem CID | 11598 |
IUPAC Name | hexa-1,5-diene |
SMILES | C=CCCC=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1365014
|
Thermo Scientific Chemicals
A1365014 |
25 g |
Each for $68.86
|
|
|||||
AAA1365022
|
Thermo Scientific Chemicals
A1365022 |
100 g |
Each for $153.40
|
|
|||||
AAA1365036
|
Thermo Scientific Chemicals
A1365036 |
500 g |
Each for $560.85
|
|
|||||
Description
1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.
Solubility
Soluble in water(0.17g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
592-42-7 | |
82.146 | |
PYGSKMBEVAICCR-UHFFFAOYSA-N | |
11598 | |
C=CCCC=C |
C6H10 | |
MFCD00008666 | |
1,5-hexadiene, biallyl, diallyl, hexadiene dot, alpha,omega-hexadiene, .alpha.,.omega.-hexadiene, unii-4mtz4764fi, 1,5-hexanediene, acmc-1axer, hexa-1,5-dien-1-yl | |
hexa-1,5-diene |
Specifications
592-42-7 | |
0.692 | |
−27°C (−16°F) | |
C6H10 | |
MFCD00008666 | |
UN2458 | |
1,5-hexadiene, biallyl, diallyl, hexadiene dot, alpha,omega-hexadiene, .alpha.,.omega.-hexadiene, unii-4mtz4764fi, 1,5-hexanediene, acmc-1axer, hexa-1,5-dien-1-yl | |
PYGSKMBEVAICCR-UHFFFAOYSA-N | |
hexa-1,5-diene | |
11598 | |
98% |
-141°C | |
58°C to 60°C | |
Turpentine-like | |
1.404 | |
25 g | |
1731106 | |
Soluble in water(0.17g/L). | |
C=CCCC=C | |
82.146 | |
82.15 | |
1,5-Hexadiene |
Safety and Handling
GHS H Statement
H225-H304-H319-H335
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P331-P332+P313-P362-P370+P378q-P501c
H225-H304-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXADIENES
EINECSNumber : 209-754-7
RTECSNumber : MM1997800
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only