missing translation for 'onlineSavingsMsg'
Learn More

1,5-Hexadiyne, 50% in pentane

CAS: 628-16-0 | C6H6 | 78.114 g/mol

$227.73 - $822.63

Chemical Identifiers

CAS 628-16-0
Molecular Formula C6H6
Molecular Weight (g/mol) 78.114
MDL Number MFCD00014924
InChI Key YFIBSNDOVCWPBL-UHFFFAOYSA-N
Synonym 1,5-hexadiyne, dipropargyl, bipropargyl, hc#cch2ch2c#ch, 1,5-hexadiyne in pentane, hccch2ch2cch, acmc-1b142, 1,5-hexadiyne 6ci, 8ci, 9ci, 1,5-hexadiyne solution, 50 wt. % in pentane 1g
PubChem CID 69402
ChEBI CHEBI:37821
IUPAC Name hexa-1,5-diyne
SMILES C#CCCC#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1872004
View Documents
Thermo Scientific Chemicals
01872004
2 g
Each for $227.73
Only null left
 
AA1872009
View Documents
Thermo Scientific Chemicals
01872009
10 g
Each for $822.63
Only null left
 
Description

Description

1,5-Hexadiyne is used in the preparation of bisdehydro[12]annulene and biphenylene. It is also used as an important starting material in the preparation of (5Z,9Z)-5,9-hexadecadienoic acid. Further, it is involved in the preparation of linear organotin polymers by reaction with organotin dihydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

628-16-0
78.114
YFIBSNDOVCWPBL-UHFFFAOYSA-N
69402
hexa-1,5-diyne
C6H6
MFCD00014924
1,5-hexadiyne, dipropargyl, bipropargyl, hc#cch2ch2c#ch, 1,5-hexadiyne in pentane, hccch2ch2cch, acmc-1b142, 1,5-hexadiyne 6ci, 8ci, 9ci, 1,5-hexadiyne solution, 50 wt. % in pentane 1g
CHEBI:37821
C#CCCC#C

Specifications

628-16-0
-2°C (29°F)
MFCD00014924
UN3295
Immiscible with water.
C#CCCC#C
78.114
CHEBI:37821
50% in pentane
1,5-Hexadiyne
0.805 g/mL
C6H6
2 g
1,5-hexadiyne, dipropargyl, bipropargyl, hc#cch2ch2c#ch, 1,5-hexadiyne in pentane, hccch2ch2cch, acmc-1b142, 1,5-hexadiyne 6ci, 8ci, 9ci, 1,5-hexadiyne solution, 50 wt. % in pentane 1g
YFIBSNDOVCWPBL-UHFFFAOYSA-N
hexa-1,5-diyne
69402
78.11
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only