missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals

$63.37 - $167.27

Chemical Identifiers

CAS 54-95-5
Molecular Formula C6H10N4
Molecular Weight (g/mol) 138.174
MDL Number MFCD00005939
InChI Key CWRVKFFCRWGWCS-UHFFFAOYSA-N
Synonym pentylenetetrazol, pentylenetetrazole, pentetrazol, metrazole, cardiazol, cardiazole, cardiol, corazol, 1,5-pentamethylenetetrazole, corazole
PubChem CID 5917
ChEBI CHEBI:34910
IUPAC Name 6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
SMILES C1CCC2=NN=NN2CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1618506
View Documents
Thermo Scientific Chemicals
L1618506
5 g
Each for $63.37
 
AAL1618514
View Documents
Thermo Scientific Chemicals
L1618514
25 g
Each for $167.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54-95-5
138.174
CWRVKFFCRWGWCS-UHFFFAOYSA-N
5917
6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
C6H10N4
MFCD00005939
pentylenetetrazol, pentylenetetrazole, pentetrazol, metrazole, cardiazol, cardiazole, cardiol, corazol, 1,5-pentamethylenetetrazole, corazole
CHEBI:34910
C1CCC2=NN=NN2CC1

Specifications

54-95-5
194°C (12 mmHg)
MFCD00005939
UN2811
Hygroscopic
pentylenetetrazol, pentylenetetrazole, pentetrazol, metrazole, cardiazol, cardiazole, cardiol, corazol, 1,5-pentamethylenetetrazole, corazole
C1CCC2=NN=NN2CC1
138.174
CHEBI:34910
98%
59°C to 61°C
C6H10N4
5 g
135492
14,7141
CWRVKFFCRWGWCS-UHFFFAOYSA-N
6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
5917
138.17
1,5-Pentamethylene-1H-tetrazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 200-219-3

RTECSNumber : XF8225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only