Learn More
1,5-Pentanediol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1399836
Description
1,5-Pentanediol is used as a plasticizer in cellulose products and adhesives. It is used as a brake fluid additive. It reacts with 3,4-dihydro-2H-pyran to get 5-tetrahydropyran-2-yloxy-pentan-1-ol. It is also used to prepare polyesters for emulsifying agents and resin intermediates. Further, it is used in ink, toner and colorant products. In addition to this, it is used in brake fluid compositions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,5-Pentanediol is used as a plasticizer in cellulose products and adhesives. It is used as a brake fluid additive. It reacts with 3,4-dihydro-2H-pyran to get 5-tetrahydropyran-2-yloxy-pentan-1-ol. It is also used to prepare polyesters for emulsifying agents and resin intermediates. Further, it is used in ink, toner and colorant products. In addition to this, it is used in brake fluid compositions.
Solubility
Miscible with water, methanol, ethanol and acetone.
Notes
Hygroscopic. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, chloroformates and reducing agents.
Specifications
1,5-Pentanediol | |
-16°C | |
241°C to 242°C | |
C5H12O2 | |
HO(CH2)5OH | |
500 g | |
Hygroscopic | |
1,5-pentanediol, 1,5-dihydroxypentane, pentylene glycol, pentamethylene glycol, 1,5-pentylene glycol, 1,5 pentanediol, 1,5-pentamethylene glycol, alpha,omega-pentanediol, unii-07uxz0scst, 1,5-pentandiol | |
ALQSHHUCVQOPAS-UHFFFAOYSA-N | |
pentane-1,5-diol | |
8105 | |
97% |
111-29-5 | |
0.992 | |
129°C (264°F) | |
1.45 | |
MFCD00002978 | |
1560130 | |
14,7117 | |
Miscible with water,methanol,ethanol and acetone. | |
OCCCCCO | |
104.15 | |
104.15 |
Chemical Identifiers
111-29-5 | |
104.15 | |
ALQSHHUCVQOPAS-UHFFFAOYSA-N | |
8105 | |
OCCCCCO |
C5H12O2 | |
MFCD00002978 | |
1,5-pentanediol, 1,5-dihydroxypentane, pentylene glycol, pentamethylene glycol, 1,5-pentylene glycol, 1,5 pentanediol, 1,5-pentamethylene glycol, alpha,omega-pentanediol, unii-07uxz0scst, 1,5-pentandiol | |
pentane-1,5-diol |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 203-854-4
RTECSNumber : SA0480000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only