missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid, Thermo Scientific™
$136.65 - $136.65
Chemical Identifiers
CAS | 950603-35-7 |
---|---|
Molecular Formula | C9H10F3N3O2S |
Molecular Weight (g/mol) | 281.25 |
MDL Number | MFCD11101440 |
InChI Key | PWQMNLMMMCREOV-UHFFFAOYSA-N |
Synonym | 1-5-trifluoromethyl-1,3,4-thiadiazol-2-yl piperidine-4-carboxylic acid |
PubChem CID | 33589561 |
IUPAC Name | 1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid |
SMILES | OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F |
Chemical Identifiers
950603-35-7 | |
281.25 | |
PWQMNLMMMCREOV-UHFFFAOYSA-N | |
33589561 | |
OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F |
C9H10F3N3O2S | |
MFCD11101440 | |
1-5-trifluoromethyl-1,3,4-thiadiazol-2-yl piperidine-4-carboxylic acid | |
1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid |
Specifications
950603-35-7 | |
C9H10F3N3O2S | |
250 mg | |
PWQMNLMMMCREOV-UHFFFAOYSA-N | |
1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid | |
33589561 | |
97% |
Amber Glass Bottle | |
MFCD11101440 | |
1-5-trifluoromethyl-1,3,4-thiadiazol-2-yl piperidine-4-carboxylic acid | |
OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F | |
281.25 | |
281.25 | |
1-[5-(trifluoromethyl)-1, 3, 4-thiadiazol-2-yl]piperidine-4-carboxylic acid |
Safety and Handling
GHS Signal Word: Warning