Learn More
1,6-Bismaleimidohexane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H58874MC
Description
A sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.
Solubility
Soluble in chloroform.
Notes
Protect from heat. Store away from oxidizing agents.
Spécifications
1,6-Bismaleimidohexane | |
144°C | |
MFCD00047122 | |
1,6-bismaleimidohexane, 1,6-dimaleimidohexane, n,n'-hexamethylenedimaleimide, 1,6-bis maleimido hexane, 1,1'-hexane-1,6-diyl bis 1h-pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1,1'-1,6-hexanediyl bis, maleimide, n,n'-hexamethylenedi, n,n'-hexamethylenebis maleimide, 1,1'-1,6-hexanediyl bis-1h-pyrrole-2,5-dione | |
PYVHLZLQVWXBDZ-UHFFFAOYSA-N | |
1-[6-(2,5-dioxopyrrol-1-yl)hexyl]pyrrole-2,5-dione | |
20992 | |
97% |
4856-87-5 | |
C14H16N2O4 | |
100 mg | |
Soluble in chloroform. | |
C1=CC(=O)N(C1=O)CCCCCCN2C(=O)C=CC2=O | |
276.292 | |
276.29 |
Identifiants chimiques
4856-87-5 | |
276.292 | |
PYVHLZLQVWXBDZ-UHFFFAOYSA-N | |
20992 | |
C1=CC(=O)N(C1=O)CCCCCCN2C(=O)C=CC2=O |
C14H16N2O4 | |
MFCD00047122 | |
1,6-bismaleimidohexane, 1,6-dimaleimidohexane, n,n'-hexamethylenedimaleimide, 1,6-bis maleimido hexane, 1,1'-hexane-1,6-diyl bis 1h-pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1,1'-1,6-hexanediyl bis, maleimide, n,n'-hexamethylenedi, n,n'-hexamethylenebis maleimide, 1,1'-1,6-hexanediyl bis-1h-pyrrole-2,5-dione | |
1-[6-(2,5-dioxopyrrol-1-yl)hexyl]pyrrole-2,5-dione |
Sécurité et manipulation
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 225-449-1
missing translation for 'rtecsNumber' : ON5520000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only