missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Diisocyanatohexane, 98+%, Thermo Scientific Chemicals

$68.70 - $211.62

Chemical Identifiers

CAS 822-06-0
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00002047
InChI Key RRAMGCGOFNQTLD-UHFFFAOYSA-N
Synonym hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester
PubChem CID 13192
ChEBI CHEBI:53578
IUPAC Name 1,6-diisocyanatohexane
SMILES O=C=NCCCCCCN=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4380318
View Documents
Thermo Scientific Chemicals
04380318
50 g
Each for $68.70
 
AA4380330
View Documents
Thermo Scientific Chemicals
04380330
250 g
Each for $211.62
 
Description

Description

1,6-Diisocyanatohexane is used as a polymerizing agent in polyurethane paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Chemical Identifiers

822-06-0
168.20
RRAMGCGOFNQTLD-UHFFFAOYSA-N
13192
1,6-diisocyanatohexane
C8H12N2O2
MFCD00002047
hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester
CHEBI:53578
O=C=NCCCCCCN=C=O

Specifications

822-06-0
1.05 g/mL
130°C (266°F)
C8H12N2O2
MFCD00002047
UN2281
Moisture sensitive
Reacts with water.
O=C=NCCCCCCN=C=O
168.20
CHEBI:53578
≥98%
1,6-Diisocyanatohexane
-67°C
255°C
Pungent
1.452
50 g
956709
hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester
RRAMGCGOFNQTLD-UHFFFAOYSA-N
1,6-diisocyanatohexane
13192
168.19
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H315-H319-H317-H335-H204
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fire or projection hazard.

P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302-H314-H317-H330-H334-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: HEXAMETHYLENE DIISOCYANATE

EINECSNumber : 212-485-8

RTECSNumber : MO1740000

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only