Learn More
1,6-Heptadien-4-ol, 97%
CAS: 2883-45-6 | C7H12O | 112.17 g/mol
$194.85 - $194.85
Chemical Identifiers
| CAS | 2883-45-6 |
|---|---|
| Molecular Formula | C7H12O |
| Molecular Weight (g/mol) | 112.17 |
| MDL Number | MFCD00008663 |
| InChI Key | UTGFOWQYZKTZTN-UHFFFAOYSA-N |
| Synonym | 1,6-heptadien-4-ol, 1,6-heptadiene-4-ol, diallylcarbinol, pubchem13740, 4-hepta-1,6-dienol, acmc-1cei5, intermediates-zcf02285, 4-hydroxy-1,6-heptadiene, 4-01-00-02249 beilstein handbook reference |
| PubChem CID | 17902 |
| IUPAC Name | hepta-1,6-dien-4-ol |
| SMILES | OC(CC=C)CC=C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC150090050
|
Thermo Scientific Chemicals
150090050 |
5 g | Glass bottle |
Each for $194.85
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2883-45-6 | |
| 112.17 | |
| UTGFOWQYZKTZTN-UHFFFAOYSA-N | |
| 17902 | |
| OC(CC=C)CC=C |
| C7H12O | |
| MFCD00008663 | |
| 1,6-heptadien-4-ol, 1,6-heptadiene-4-ol, diallylcarbinol, pubchem13740, 4-hepta-1,6-dienol, acmc-1cei5, intermediates-zcf02285, 4-hydroxy-1,6-heptadiene, 4-01-00-02249 beilstein handbook reference | |
| hepta-1,6-dien-4-ol |
Specifications
| 2883-45-6 | |
| 100.0 | |
| 0.8600g/mL | |
| 40°C | |
| 96% min. (GC) | |
| C7H12O | |
| H2C=CHCH2CH(OH)CH2CH=CH2 | |
| 5 g | |
| 0.86 | |
| UTGFOWQYZKTZTN-UHFFFAOYSA-N | |
| hepta-1,6-dien-4-ol | |
| 17902 | |
| 97% | |
| 1, 6-Heptadien-4-ol, 97% |
| 96.0 | |
| Colorless to Yellow | |
| 151°C | |
| Authentic | |
| Glass bottle | |
| 1.4490 to 1.453 | |
| MFCD00008663 | |
| 01, 455 | |
| 1,6-heptadien-4-ol, 1,6-heptadiene-4-ol, diallylcarbinol, pubchem13740, 4-hepta-1,6-dienol, acmc-1cei5, intermediates-zcf02285, 4-hydroxy-1,6-heptadiene, 4-01-00-02249 beilstein handbook reference | |
| OC(CC=C)CC=C | |
| 112.17 | |
| 112.17 | |
| Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
GHS Signal Word: Warning
EINECSNumber : 220-742-0
RTECSNumber : MI5450000
TSCA : TSCA
RUO – Research Use Only