missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Hexanediamine, 99.5+%, Thermo Scientific Chemicals

$58.53 - $1097.02

Chemical Identifiers

CAS 124-09-4
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
InChI Key NAQMVNRVTILPCV-UHFFFAOYSA-N
Synonym 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
PubChem CID 16402
ChEBI CHEBI:39618
IUPAC Name hexane-1,6-diamine
SMILES C(CCCN)CCN
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120641000
SDS View Documents
Thermo Scientific Chemicals
120641000
100 g Plastic bottle
Each for $58.53
 
AC120645000
SDS View Documents
Thermo Scientific Chemicals
120645000
500 g Plastic bottle
Each for $135.12
 
AC120640010
SDS View Documents
Thermo Scientific Chemicals
120640010
1 kg Plastic bottle
Each for $223.57
 
AC120640051
SDS View Documents
Thermo Scientific Chemicals
120640051
5 kg Plastic drum
Each for $1,097.02
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

124-09-4
116.21
1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
CHEBI:39618
C(CCCN)CCN
C6H16N2
NAQMVNRVTILPCV-UHFFFAOYSA-N
16402
hexane-1,6-diamine

Specifications

124-09-4
White
81°C
99.5% min. (GC)
C6H16N2
100 g
15,4727
Solubility in water: 490g/L (20°C). Other solubilities: soluble in ethanol, benzene, diethyl ether
C(CCCN)CCN
116.21
CHEBI:39618
99.5+%
1, 6-Hexanediamine
38.0°C to 41.0°C
204.0°C to 205.0°C
Authentic
Plastic bottle
H2N(CH2)6NH2
04,269
1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
NAQMVNRVTILPCV-UHFFFAOYSA-N
hexane-1,6-diamine
16402
116.21
Crystalline Low Melting Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with wate

GHS Signal Word: Danger

EINECSNumber : 204-679-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only