missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediol diacrylate, 99% (reactive esters), stab. with 90ppm hydroquinone

CAS: 13048-33-4 | C12H18O4 | 226.27 g/mol

$84.90 - $310.70

Chemical Identifiers

CAS 13048-33-4
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD00008631
InChI Key FIHBHSQYSYVZQE-UHFFFAOYSA-N
Synonym 1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate
PubChem CID 25644
IUPAC Name 6-prop-2-enoyloxyhexyl prop-2-enoate
SMILES C=CC(=O)OCCCCCCOC(=O)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4320318
View Documents
Thermo Scientific Chemicals
04320318
50 g
Each for $84.90
Only null left
 
AA4320330
View Documents
Thermo Scientific Chemicals
04320330
250 g
Each for $310.70
Only null left
 
Description

Description

1,6-Hexanediol diacrylate is used as a functional monomer for polymers. It acts as a cross linking agent between the molecular chains of polymers. Further, it is used in adhesives, sealants, alkyd coatings, elastomers, photopolymers, and inks for improved adhesion, hardness, abrasion and heat resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13048-33-4
226.27
FIHBHSQYSYVZQE-UHFFFAOYSA-N
25644
C=CC(=O)OCCCCCCOC(=O)C=C
C12H18O4
MFCD00008631
1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate
6-prop-2-enoyloxyhexyl prop-2-enoate

Specifications

13048-33-4
>110°C (>230°F)
MFCD00008631
Hygroscopic
Slightly miscible with water.
C=CC(=O)OCCCCCCOC(=O)C=C
226.27
226.28
Liquid
(reactive esters)
1.010 g/mL
C12H18O4
50 g
1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate
FIHBHSQYSYVZQE-UHFFFAOYSA-N
6-prop-2-enoyloxyhexyl prop-2-enoate
25644
99%
1,6-Hexanediol diacrylate, Stabilized with 90ppm hydroquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c

H315-H317-H319

EINECSNumber : 235-921-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only