missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,8-Diaminooctane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 112310250

 View more versions of this product

Catalog No. AC112310250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 8-Diaminooctane
97.5
50.0°C to 54.0°C
165g/mL
115°C
97.5% min. (GC)
C8H22Cl2N2
MFCD00008248
04, 271
Solubility in water: 575g/L (20°C).
[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]
217.18
CHEBI:73112
98%
373-44-4
100.0
White to Yellow
225.0°C to 226.0°C
Authentic
Glass bottle
H2N(CH2)8NH2
25 g
1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392
ZFLWZOGXFQNIMT-UHFFFAOYSA-N
octane-1,8-bis(aminium) dichloride
24250
144.26
Solidified Crystalline Mass

Chemical Identifiers

373-44-4
217.18
ZFLWZOGXFQNIMT-UHFFFAOYSA-N
24250
[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]
C8H22Cl2N2
MFCD00008248
1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392
CHEBI:73112
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physician.
Wear

GHS Signal Word: Danger

EINECSNumber : 206-764-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only