1,8-Dibromonaphthalene 98.0+%, TCI America™
Supplier:  TCI America D46565G
 View more versions of this product
Catalog No. D46565G
Explore available promotions
Description
Description
Specifications
Specifications
1,8-Dibromonaphthalene
110°C
C10H6Br2
5 g
C1=CC2=C(C(=C1)Br)C(=CC=C2)Br
285.966
285.97
Crystalline Powder
17135-74-9
140°C
MFCD00183574
DLXBGTIGAIESIG-UHFFFAOYSA-N
1,8-dibromonaphthalene
625356
≥98.0% (GC)
Chemical Identifiers
17135-74-9
285.966
DLXBGTIGAIESIG-UHFFFAOYSA-N
1,8-dibromonaphthalene
C10H6Br2
MFCD00183574
625356
C1=CC2=C(C(=C1)Br)C(=CC=C2)Br
Videos
Safety and Handling
Safety and Handling
TSCA : No