missing translation for 'onlineSavingsMsg'
Learn More

1,8-Naphthyridine 98.0+%, TCI America™

$46.60 - $272.16

Chemical Identifiers

CAS 254-60-4
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00059751
InChI Key FLBAYUMRQUHISI-UHFFFAOYSA-N
Synonym 1,8-Diazanaphthalene, Pyrido[2,3-b]pyridine
PubChem CID 136069
ChEBI CHEBI:36628
IUPAC Name 1,8-naphthyridine
SMILES C1=CC2=C(N=C1)N=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0464100MG
View Documents
TCI America
N0464100MG
100 mg N/A
Only null left
 
N04641G
View Documents
TCI America
N04641G
1 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

254-60-4
130.15
FLBAYUMRQUHISI-UHFFFAOYSA-N
136069
1,8-naphthyridine
C8H6N2
MFCD00059751
1,8-Diazanaphthalene, Pyrido[2,3-b]pyridine
CHEBI:36628
C1=CC2=C(N=C1)N=CC=C2

Specifications

254-60-4
C8H6N2
100 mg
FLBAYUMRQUHISI-UHFFFAOYSA-N
1,8-naphthyridine
136069
130.15
Crystalline Powder
98°C
MFCD00059751
1,8-Diazanaphthalene, Pyrido[2,3-b]pyridine
C1=CC2=C(N=C1)N=CC=C2
130.15
CHEBI:36628
≥98.0% (GC)
1,8-Naphthyridine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'