missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Acetylpiperidine-4-carbonyl chloride, 97%, may contain up to ca 1M free HCl, Thermo Scientific Chemicals

$152.48 - $546.03

Chemical Identifiers

CAS 59084-16-1
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.639
MDL Number MFCD02094017
InChI Key OHCPVLJEAHBMEG-UHFFFAOYSA-N
Synonym 1-acetylisonipecotoyl chloride, n-acetylisonipecotoyl chloride, 1-acetyl-4-piperidinecarbonyl chloride, 1-acetyl isonipecotoyl chloride, acmc-20aoys, acetylisonipecotoyl chloride, 1acetylisonipecotoyl chloride, n-acetylisonipecotyl chloride, ksc274a6b, n-acetyl isonipecotoyl chloride
PubChem CID 100950
IUPAC Name 1-acetylpiperidine-4-carbonyl chloride
SMILES CC(=O)N1CCC(CC1)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1842603
View Documents
Thermo Scientific Chemicals
L1842603
1 g
Each for $152.48
 
AAL1842606
View Documents
Thermo Scientific Chemicals
L1842606
5 g
Each for $546.03
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

59084-16-1
189.639
OHCPVLJEAHBMEG-UHFFFAOYSA-N
100950
CC(=O)N1CCC(CC1)C(=O)Cl
C8H12ClNO2
MFCD02094017
1-acetylisonipecotoyl chloride, n-acetylisonipecotoyl chloride, 1-acetyl-4-piperidinecarbonyl chloride, 1-acetyl isonipecotoyl chloride, acmc-20aoys, acetylisonipecotoyl chloride, 1acetylisonipecotoyl chloride, n-acetylisonipecotyl chloride, ksc274a6b, n-acetyl isonipecotoyl chloride
1-acetylpiperidine-4-carbonyl chloride

Specifications

59084-16-1
C8H12ClNO2
1 g
389571
1-acetylisonipecotoyl chloride, n-acetylisonipecotoyl chloride, 1-acetyl-4-piperidinecarbonyl chloride, 1-acetyl isonipecotoyl chloride, acmc-20aoys, acetylisonipecotoyl chloride, 1acetylisonipecotoyl chloride, n-acetylisonipecotyl chloride, ksc274a6b, n-acetyl isonipecotoyl chloride
CC(=O)N1CCC(CC1)C(=O)Cl
189.639
189.64
1-Acetylpiperidine-4-carbonyl chloride, may contain up to ≈1M free HCl
131°C to 133°C
MFCD02094017
UN3261
Moisture sensitive
OHCPVLJEAHBMEG-UHFFFAOYSA-N
1-acetylpiperidine-4-carbonyl chloride
100950
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 261-594-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only