missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Allyl-2-thiourea 98.0+%, TCI America™

$80.97 - $665.57

Chemical Identifiers

CAS 109-57-9
Molecular Formula C4H8N2S
Molecular Weight (g/mol) 116.182
MDL Number MFCD00004940
InChI Key HTKFORQRBXIQHD-UHFFFAOYSA-N
Synonym allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
PubChem CID 1549517
ChEBI CHEBI:74079
IUPAC Name prop-2-enylthiourea
SMILES C=CCNC(=S)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A022025G
View Documents
TCI America
A022025G
25 g
Each for $80.97
 
A0220100G
View Documents
TCI America
A0220100G
100 g
Each for $199.48
 
A0220500G
View Documents
TCI America
A0220500G
500 g
Each for $665.57
 
Description

Description

Specifications

Chemical Identifiers

109-57-9
116.182
HTKFORQRBXIQHD-UHFFFAOYSA-N
1549517
prop-2-enylthiourea
C4H8N2S
MFCD00004940
allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
CHEBI:74079
C=CCNC(=S)N

Specifications

109-57-9
White-Yellow
MFCD00004940
2811
HTKFORQRBXIQHD-UHFFFAOYSA-N
prop-2-enylthiourea
1549517
116.18
Crystalline Powder
71°C
C4H8N2S
25 g
allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
C=CCNC(=S)N
116.182
CHEBI:74079
≥98.0% (T)
1-Allyl-2-thiourea
Videos
Safety and Handling

Safety and Handling

RTECSNumber : YR8050000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'