missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate 96.0+%, TCI America™
$77.40 - $307.94
Chemical Identifiers
CAS | 144120-54-7 |
---|---|
Molecular Formula | C23H23NO6 |
Molecular Weight (g/mol) | 409.44 |
MDL Number | MFCD00467718 |
InChI Key | ORKKMGRINLTBPC-UHFFFAOYNA-N |
Synonym | fmoc-glu-oall, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester, fmoc-l-glutamic acid 1-allyl ester, fmoc-l-glutamic acid, fmoc-l-glutamic acid, fmoc-glu-allyl ester, fmoc-glu-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate |
PubChem CID | 7020606 |
IUPAC Name | 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid |
SMILES | OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C |
Chemical Identifiers
144120-54-7 | |
409.44 | |
ORKKMGRINLTBPC-UHFFFAOYNA-N | |
7020606 | |
OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C |
C23H23NO6 | |
MFCD00467718 | |
fmoc-glu-oall, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester, fmoc-l-glutamic acid 1-allyl ester, fmoc-l-glutamic acid, fmoc-l-glutamic acid, fmoc-glu-allyl ester, fmoc-glu-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate | |
4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid |
Specifications
144120-54-7 | |
White-Yellow | |
MFCD00467718 | |
fmoc-glu-oall, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester, fmoc-l-glutamic acid 1-allyl ester, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, fmoc-l-glutamic acid, fmoc-glu-allyl ester, fmoc-glu-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, fmoc-glu-oall hplc, fmoc-glu-oall hplc, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate | |
OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C | |
409.44 | |
409.44 | |
Crystalline Powder |
118°C | |
C23H23NO6 | |
1 g | |
ORKKMGRINLTBPC-UHFFFAOYNA-N | |
4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid | |
7020606 | |
≥96.0% (HPLC,T) | |
1-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator