missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Allylnaphthalene 98.0+%, TCI America™
Supplier: TCI America A24655G
Specifications
1-Allylnaphthalene | |
Yellow | |
C13H12 | |
5 g | |
RJFCFNWLPJRCLR-UHFFFAOYSA-N | |
1-(prop-2-en-1-yl)naphthalene | |
17217 | |
≥98.0% (GC) |
2489-86-3 | |
133°C | |
MFCD00021581 | |
3-(1-Naphthyl)-1-propene | |
C=CCC1=C2C=CC=CC2=CC=C1 | |
168.24 | |
168.24 | |
Liquid |
Safety and Handling
RTECSNumber : QJ1399900
TSCA : No