missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-2-methylpyridinium Iodide 98.0+%, TCI America™

$178.95 - $605.83

Chemical Identifiers

CAS 7583-90-6
Molecular Formula C6H9IN2
Molecular Weight (g/mol) 236.056
MDL Number MFCD00955862
InChI Key XMFXSCPQGJZSLU-UHFFFAOYSA-M
PubChem CID 12291507
IUPAC Name 2-methylpyridin-1-ium-1-amine;iodide
SMILES CC1=CC=CC=[N+]1N.[I-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A25581G
View Documents
TCI America
A25581G
1 g
Each for $178.95
Only null left
 
A25585G
View Documents
TCI America
A25585G
5 g
Each for $605.83
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7583-90-6
236.056
XMFXSCPQGJZSLU-UHFFFAOYSA-M
2-methylpyridin-1-ium-1-amine;iodide
C6H9IN2
MFCD00955862
12291507
CC1=CC=CC=[N+]1N.[I-]

Specifications

7583-90-6
White-Yellow
MFCD00955862
XMFXSCPQGJZSLU-UHFFFAOYSA-M
2-methylpyridin-1-ium-1-amine;iodide
12291507
≥98.0% (HPLC,T)
1-Amino-2-methylpyridinium Iodide
152°C
C6H9IN2
1 g
CC1=CC=CC=[N+]1N.[I-]
236.056
236.06
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'