missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic Acid 95.0+%, TCI America™

$329.94 - $963.71

Chemical Identifiers

CAS 905954-28-1
Molecular Formula C15H31NO8
Molecular Weight (g/mol) 353.412
InChI Key PVRGRRPCHFTMMD-UHFFFAOYSA-N
Synonym 21-Amino-4,7,10,13,16,19-hexaoxahenicosanoic Acid, Amino-PEG6-carboxylic Acid, Carboxy-PEG6-amine
PubChem CID 51035062
IUPAC Name 3-[2-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]propanoic acid
SMILES C(COCCOCCOCCOCCOCCOCCN)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A2526100MG
View Documents
TCI America
A2526100MG
100 mg
Each for $329.94
Only null left
 
A2526500MG
View Documents
TCI America
A2526500MG
500 mg
Each for $963.71
Only null left
 
Description

Description

Specifications

Chemical Identifiers

905954-28-1
353.412
21-Amino-4,7,10,13,16,19-hexaoxahenicosanoic Acid, Amino-PEG6-carboxylic Acid, Carboxy-PEG6-amine
3-[2-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]propanoic acid
C15H31NO8
PVRGRRPCHFTMMD-UHFFFAOYSA-N
51035062
C(COCCOCCOCCOCCOCCOCCN)C(=O)O

Specifications

905954-28-1
C15H31NO8
PVRGRRPCHFTMMD-UHFFFAOYSA-N
3-[2-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]propanoic acid
353.412
353.41
Crystalline Powder
White-Yellow
21-Amino-4,7,10,13,16,19-hexaoxahenicosanoic Acid, Amino-PEG6-carboxylic Acid, Carboxy-PEG6-amine
C(COCCOCCOCCOCCOCCOCCN)C(=O)O
100 mg
51035062
≥95.0% (HPLC)
1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'