Learn More
1-Amino-3,6,9,12-tetraoxapentadec-14-yne, Thermo Scientific Chemicals
Acetylene containing reagent with PEG spacer and carboxyl reactivity
$701.56 - $1761.89
Chemical Identifiers
CAS | 1013921-36-2 |
---|---|
Molecular Formula | C11H21NO4 |
Molecular Weight (g/mol) | 231.292 |
MDL Number | MFCD22380736 |
InChI Key | QDLPAHLHHBCWOW-UHFFFAOYSA-N |
Synonym | 3,6,9,12-tetraoxapentadec-14-yn-1-amine, amino-peg4-alkyne, propyne-peg4-amine, h2n-peg4-propyne, propyne-peg4-nh2, propargyl-peg4-amine, 1-amino-3,6,9,12-tetraoxapentadec-14-yne, hc inverted exclamation markoc-ch2-peg4-nh2, 2-2-2-2-propargyloxy ethoxy ethoxy ethoxy ethylamine |
PubChem CID | 86580391 |
IUPAC Name | 2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethanamine |
SMILES | C#CCOCCOCCOCCOCCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ64527MC
|
Thermo Scientific Chemicals
J64527MC |
100 mg |
Each for $701.56
|
|
|||||
AAJ64527ME
|
Thermo Scientific Chemicals
J64527ME |
500 mg |
Each for $1,761.89
|
|
|||||
Description
1-Amino-3,6,9,12-tetraoxapentadec-14-yne is used as a acetylene containing reagent with PEG spacer and carboxyl reactivity. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification of carboxylic acid containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Amino-3,6,9,12-tetraoxapentadec-14-yne is used as a acetylene containing reagent with PEG spacer and carboxyl reactivity. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification of carboxylic acid containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character.
Solubility
Soluble in DMSO, DMF, DCM, THF, or chloroform.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers..
Chemical Identifiers
1013921-36-2 | |
231.292 | |
QDLPAHLHHBCWOW-UHFFFAOYSA-N | |
86580391 | |
C#CCOCCOCCOCCOCCN |
C11H21NO4 | |
MFCD22380736 | |
3,6,9,12-tetraoxapentadec-14-yn-1-amine, amino-peg4-alkyne, propyne-peg4-amine, h2n-peg4-propyne, propyne-peg4-nh2, propargyl-peg4-amine, 1-amino-3,6,9,12-tetraoxapentadec-14-yne, hc inverted exclamation markoc-ch2-peg4-nh2, 2-2-2-2-propargyloxy ethoxy ethoxy ethoxy ethylamine | |
2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethanamine |
Specifications
1013921-36-2 | |
C11H21NO4 | |
MFCD22380736 | |
3,6,9,12-tetraoxapentadec-14-yn-1-amine, amino-peg4-alkyne, propyne-peg4-amine, h2n-peg4-propyne, propyne-peg4-nh2, propargyl-peg4-amine, 1-amino-3,6,9,12-tetraoxapentadec-14-yne, hc inverted exclamation markoc-ch2-peg4-nh2, 2-2-2-2-propargyloxy ethoxy ethoxy ethoxy ethylamine | |
QDLPAHLHHBCWOW-UHFFFAOYSA-N | |
2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethanamine | |
86580391 | |
Solid |
Yellow | |
NH2CH2CH2(OCH2CH2)3OCH2C≡CH | |
100 mg | |
Soluble in DMSO,DMF,DCM,THF,or chloroform. | |
C#CCOCCOCCOCCOCCN | |
231.292 | |
231.29 | |
1-Amino-3,6,9,12-tetraoxapentadec-14-yne |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only