missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzothiophen-5-amine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 20532-28-9
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
InChI Key ZUPYTANKWDPRDP-UHFFFAOYSA-N
Synonym benzo b thiophen-5-amine, 5-aminobenzothiophene, 5-aminobenzo b thiophene, benzo b thiophen-5-ylamine, 5-aminothionaphthene, benzothiophen-5-amine, 1-benzothien-5-ylamine, 5-amino-benzo b thiophene, benzo b thiophene-5-ylamine
PubChem CID 288032
IUPAC Name 1-benzothiophen-5-amine
SMILES C1=CC2=C(C=CS2)C=C1N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC24814CB
SDS View Documents
Thermo Scientific Chemicals
CC24814CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC24814DA
SDS View Documents
Thermo Scientific Chemicals
CC24814DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC24814DE
SDS View Documents
Thermo Scientific Chemicals
CC24814DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

20532-28-9
149.211
benzo b thiophen-5-amine, 5-aminobenzothiophene, 5-aminobenzo b thiophene, benzo b thiophen-5-ylamine, 5-aminothionaphthene, benzothiophen-5-amine, 1-benzothien-5-ylamine, 5-amino-benzo b thiophene, benzo b thiophene-5-ylamine
1-benzothiophen-5-amine
C8H7NS
ZUPYTANKWDPRDP-UHFFFAOYSA-N
288032
C1=CC2=C(C=CS2)C=C1N

Specifications

20532-28-9
Amber Glass Bottle
250mg
ZUPYTANKWDPRDP-UHFFFAOYSA-N
1-benzothiophen-5-amine
288032
97%
97%
C8H7NS
benzo b thiophen-5-amine, 5-aminobenzothiophene, 5-aminobenzo b thiophene, benzo b thiophen-5-ylamine, 5-aminothionaphthene, benzothiophen-5-amine, 1-benzothien-5-ylamine, 5-amino-benzo b thiophene, benzo b thiophene-5-ylamine
C1=CC2=C(C=CS2)C=C1N
149.211
149.22
1-Benzothiophen-5-amine
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'