missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl-1,4-dihydronicotinamide 95.0+%, TCI America™
$121.16 - $1002.86
Chemical Identifiers
| CAS | 952-92-1 |
|---|---|
| Molecular Formula | C13H14N2O |
| Molecular Weight (g/mol) | 214.268 |
| MDL Number | MFCD00059769 |
| InChI Key | CMNUYDSETOTBDE-UHFFFAOYSA-N |
| PubChem CID | 95276 |
| IUPAC Name | 1-benzyl-4H-pyridine-3-carboxamide |
| SMILES | C1C=CN(C=C1C(=O)N)CC2=CC=CC=C2 |
Chemical Identifiers
| 952-92-1 | |
| 214.268 | |
| CMNUYDSETOTBDE-UHFFFAOYSA-N | |
| 1-benzyl-4H-pyridine-3-carboxamide |
| C13H14N2O | |
| MFCD00059769 | |
| 95276 | |
| C1C=CN(C=C1C(=O)N)CC2=CC=CC=C2 |
Specifications
| 952-92-1 | |
| C13H14N2O | |
| 1 g | |
| C1C=CN(C=C1C(=O)N)CC2=CC=CC=C2 | |
| 214.268 | |
| 214.27 | |
| Crystalline Powder |
| 121°C | |
| MFCD00059769 | |
| CMNUYDSETOTBDE-UHFFFAOYSA-N | |
| 1-benzyl-4H-pyridine-3-carboxamide | |
| 95276 | |
| ≥95.0% (HPLC,N) | |
| 1-Benzyl-1,4-dihydronicotinamide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator