missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Benzyl-3-oxopiperazine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H55072MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Benzyl-3-oxopiperazine | |
148°C to 155°C | |
MFCD06795962 | |
1-benzyl-3-oxopiperazine, 4-benzyl-piperazin-2-one, piperazinone, 4-phenylmethyl, 2-piperazinone,4-phenylmethyl, 4-benzyl-2-piperazinone, 4-benzylpiperazine-2-one, 4-phenylmethyl piperazinone, aronis25032 | |
C1CN(CC(=O)N1)CC2=CC=CC=C2 | |
190.246 | |
190.24 |
13754-41-1 | |
C11H14N2O | |
250 mg | |
SBWVHKNCFZRBRJ-UHFFFAOYSA-N | |
4-benzylpiperazin-2-one | |
11458177 | |
95% |
Chemical Identifiers
13754-41-1 | |
190.246 | |
SBWVHKNCFZRBRJ-UHFFFAOYSA-N | |
11458177 | |
C1CN(CC(=O)N1)CC2=CC=CC=C2 |
C11H14N2O | |
MFCD06795962 | |
1-benzyl-3-oxopiperazine, 4-benzyl-piperazin-2-one, piperazinone, 4-phenylmethyl, 2-piperazinone,4-phenylmethyl, 4-benzyl-2-piperazinone, 4-benzylpiperazine-2-one, 4-phenylmethyl piperazinone, aronis25032 | |
4-benzylpiperazin-2-one |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures