missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-[(Benzyloxy)carbonyl]-2-indolinecarboxylic acid, ≥90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC13401DA
Specifications
1-[(Benzyloxy)carbonyl]-2-indolinecarboxylic acid | |
89.5 | |
90%+ | |
C17H15NO4 | |
1g | |
BSOYWTITVKXHLM-UHFFFAOYNA-N | |
1-phenylmethoxycarbonyl-2,3-dihydroindole-2-carboxylic acid | |
2776400 | |
90% |
117483-89-3 | |
100.0 | |
Amber glass bottle | |
MFCD02682032 | |
1-benzyloxy carbonyl-2-indolinecarboxylic acid, 1-benzyloxy carbonyl indoline-2-carboxylic acid, 1-benzyloxy carbonyl-2,3-dihydroindole-2-carboxylic acid, 2,3-dihydro-1h-indole-1,2-dicarboxylic acid 1-benzyl ester, 1-benzyloxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, acmc-1brpx, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, 2s, 1-benzyloxycarbonyl-2-indoline carboxylic acid, 1-benzyl-oxy carbonyl-2-indolinecarboxylic acid | |
OC(=O)C1CC2=CC=CC=C2N1C(=O)OCC1=CC=CC=C1 | |
297.31 | |
297.31 |
Chemical Identifiers
117483-89-3 | |
297.31 | |
BSOYWTITVKXHLM-UHFFFAOYNA-N | |
2776400 | |
OC(=O)C1CC2=CC=CC=C2N1C(=O)OCC1=CC=CC=C1 |
C17H15NO4 | |
MFCD02682032 | |
1-benzyloxy carbonyl-2-indolinecarboxylic acid, 1-benzyloxy carbonyl indoline-2-carboxylic acid, 1-benzyloxy carbonyl-2,3-dihydroindole-2-carboxylic acid, 2,3-dihydro-1h-indole-1,2-dicarboxylic acid 1-benzyl ester, 1-benzyloxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, acmc-1brpx, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, 2s, 1-benzyloxycarbonyl-2-indoline carboxylic acid, 1-benzyl-oxy carbonyl-2-indolinecarboxylic acid | |
1-phenylmethoxycarbonyl-2,3-dihydroindole-2-carboxylic acid |