missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[(Benzyloxy)carbonyl]-2-indolinecarboxylic acid, ≥90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC13401DE

 View more versions of this product

Catalog No. CC13401DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-[(Benzyloxy)carbonyl]-2-indolinecarboxylic acid
89.5
90%+
MFCD02682032
1-benzyloxy carbonyl-2-indolinecarboxylic acid, 1-benzyloxy carbonyl indoline-2-carboxylic acid, 1-benzyloxy carbonyl-2,3-dihydroindole-2-carboxylic acid, 2,3-dihydro-1h-indole-1,2-dicarboxylic acid 1-benzyl ester, 1-benzyloxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, acmc-1brpx, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, 2s, 1-benzyloxycarbonyl-2-indoline carboxylic acid, 1-benzyl-oxy carbonyl-2-indolinecarboxylic acid
OC(=O)C1CC2=CC=CC=C2N1C(=O)OCC1=CC=CC=C1
297.31
297.31
117483-89-3
100.0
C17H15NO4
5g
BSOYWTITVKXHLM-UHFFFAOYNA-N
1-phenylmethoxycarbonyl-2,3-dihydroindole-2-carboxylic acid
2776400
90%

Chemical Identifiers

117483-89-3
297.31
BSOYWTITVKXHLM-UHFFFAOYNA-N
2776400
OC(=O)C1CC2=CC=CC=C2N1C(=O)OCC1=CC=CC=C1
C17H15NO4
MFCD02682032
1-benzyloxy carbonyl-2-indolinecarboxylic acid, 1-benzyloxy carbonyl indoline-2-carboxylic acid, 1-benzyloxy carbonyl-2,3-dihydroindole-2-carboxylic acid, 2,3-dihydro-1h-indole-1,2-dicarboxylic acid 1-benzyl ester, 1-benzyloxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, acmc-1brpx, 1h-indole-1,2-dicarboxylicacid, 2,3-dihydro-, 1-phenylmethyl ester, 2s, 1-benzyloxycarbonyl-2-indoline carboxylic acid, 1-benzyl-oxy carbonyl-2-indolinecarboxylic acid
1-phenylmethoxycarbonyl-2,3-dihydroindole-2-carboxylic acid
Videos
SDS Documents

Documents

Promotions

Promotions