missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-1-Boc-3-hydroxypyrrolidine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2875006

 View more versions of this product

Catalog No. AAH2875006

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(+/-)-1-Boc-3-hydroxypyrrolidine
60°C to 62°C
MFCD04038535
1-boc-3-hydroxypyrrolidine, 1-boc-3-pyrrolidinol, n-boc-3-hydroxypyrrolidine, n-boc-3-pyrrolidinol, 1-n-boc-3-hydroxypyrrolidine, 1-pyrrolidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-pyrrolidinol, 3-hydroxy-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-hydroxypyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-hydroxypyrrolidine
CC(C)(C)OC(=O)N1CCC(C1)O
187.239
187.24
103057-44-9
C9H17NO3
5 g
APCBTRDHCDOPNY-UHFFFAOYSA-N
tert-butyl 3-hydroxypyrrolidine-1-carboxylate
4416939
97%

Chemical Identifiers

103057-44-9
187.239
APCBTRDHCDOPNY-UHFFFAOYSA-N
4416939
CC(C)(C)OC(=O)N1CCC(C1)O
C9H17NO3
MFCD04038535
1-boc-3-hydroxypyrrolidine, 1-boc-3-pyrrolidinol, n-boc-3-hydroxypyrrolidine, n-boc-3-pyrrolidinol, 1-n-boc-3-hydroxypyrrolidine, 1-pyrrolidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-pyrrolidinol, 3-hydroxy-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-hydroxypyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-hydroxypyrrolidine
tert-butyl 3-hydroxypyrrolidine-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only