Learn More
1-Boc-4-(4-aminophenyl)piperazine, 97%, Thermo Scientific Chemicals
$162.14 - $691.51
Chemical Identifiers
CAS | 170911-92-9 |
---|---|
Molecular Formula | C15H23N3O2 |
Molecular Weight (g/mol) | 277.368 |
MDL Number | MFCD04115065 |
InChI Key | RXFHRKPNLPBDGE-UHFFFAOYSA-N |
Synonym | tert-butyl 4-4-aminophenyl piperazine-1-carboxylate, 4-4-aminophenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-piperazin-1-yl aniline, 1-boc-4-4-aminophenyl piperazine, 1-piperazinecarboxylic acid, 4-4-aminophenyl-, 1,1-dimethylethyl ester, 4-4-aminophenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-boc-4-4'-aminophenyl piperazine dihydrochloride, 1-boc-4-4-aminophenyl-piperazine, pubchem12198 |
PubChem CID | 11011301 |
IUPAC Name | tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6319103
|
Thermo Scientific Chemicals
H6319103 |
1 g |
N/A
|
|
|||||
AAH6319106
|
Thermo Scientific Chemicals
H6319106 |
5 g |
N/A
|
|
|||||
Description
1-Boc-4-(4-aminophenyl)piperazine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Boc-4-(4-aminophenyl)piperazine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
170911-92-9 | |
277.368 | |
RXFHRKPNLPBDGE-UHFFFAOYSA-N | |
11011301 | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)N |
C15H23N3O2 | |
MFCD04115065 | |
tert-butyl 4-4-aminophenyl piperazine-1-carboxylate, 4-4-aminophenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-piperazin-1-yl aniline, 1-boc-4-4-aminophenyl piperazine, 1-piperazinecarboxylic acid, 4-4-aminophenyl-, 1,1-dimethylethyl ester, 4-4-aminophenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-boc-4-4'-aminophenyl piperazine dihydrochloride, 1-boc-4-4-aminophenyl-piperazine, pubchem12198 | |
tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate |
Specifications
170911-92-9 | |
MFCD04115065 | |
tert-butyl 4-4-aminophenyl piperazine-1-carboxylate, 4-4-aminophenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-piperazin-1-yl aniline, 1-boc-4-4-aminophenyl piperazine, 1-piperazinecarboxylic acid, 4-4-aminophenyl-, 1,1-dimethylethyl ester, 4-4-aminophenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-boc-4-4'-aminophenyl piperazine dihydrochloride, 1-boc-4-4-aminophenyl-piperazine, pubchem12198 | |
RXFHRKPNLPBDGE-UHFFFAOYSA-N | |
tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate | |
11011301 | |
97% | |
1-Boc-4-(4-aminophenyl)piperazine |
C15H23N3O2 | |
1 g | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)N | |
277.368 | |
277.36 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only