missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Boc-4-hydroxypiperidine, 98%, Thermo Scientific Chemicals

$122.74 - $122.74

Chemical Identifiers

CAS 109384-19-2
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD01075174
InChI Key PWQLFIKTGRINFF-UHFFFAOYSA-N
Synonym n-boc-4-hydroxypiperidine, 1-boc-4-hydroxypiperidine, n-boc-4-piperidinol, 1-boc-4-piperidinol, n-boc-4-hydroxylpiperidine, n-boc-4-hydroxy piperidine, 1-tert-butoxycarbonyl-4-hydroxypiperidine, 4-hydroxy-n-boc-piperidine, 1-boc-4-hydroxy-piperidine
PubChem CID 643502
IUPAC Name tert-butyl 4-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1927506
View Documents
Thermo Scientific Chemicals
L1927506
5 g
Each for $122.74
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109384-19-2
201.266
PWQLFIKTGRINFF-UHFFFAOYSA-N
643502
CC(C)(C)OC(=O)N1CCC(CC1)O
C10H19NO3
MFCD01075174
n-boc-4-hydroxypiperidine, 1-boc-4-hydroxypiperidine, n-boc-4-piperidinol, 1-boc-4-piperidinol, n-boc-4-hydroxylpiperidine, n-boc-4-hydroxy piperidine, 1-tert-butoxycarbonyl-4-hydroxypiperidine, 4-hydroxy-n-boc-piperidine, 1-boc-4-hydroxy-piperidine
tert-butyl 4-hydroxypiperidine-1-carboxylate

Specifications

109384-19-2
C10H19NO3
5 g
n-boc-4-hydroxypiperidine, 1-boc-4-hydroxypiperidine, n-boc-4-piperidinol, 1-boc-4-piperidinol, n-boc-4-hydroxylpiperidine, n-boc-4-hydroxy piperidine, 1-tert-butoxycarbonyl-4-hydroxypiperidine, 4-hydroxy-n-boc-piperidine, 1-boc-4-hydroxy-piperidine
CC(C)(C)OC(=O)N1CCC(CC1)O
201.266
201.27
1-Boc-4-hydroxypiperidine
62°C to 64°C
MFCD01075174
7202094
PWQLFIKTGRINFF-UHFFFAOYSA-N
tert-butyl 4-hydroxypiperidine-1-carboxylate
643502
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only