missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Boc-5-(tert-butyldimethylsiloxy)indole-2-boronic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H52511MD

 View more versions of this product

Catalog No. AAH52511MD



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Boc-5-(tert-butyldimethylsiloxy)indole-2-boronic acid
104°C to 106°C
MFCD08689532
5-tert-butyldimethylsilyloxy-1h-indole-2-boronic acid, n-boc protected, 1-boc-5-tbdmso-indole-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy-1h-indol-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy-1h-indol-2-ylboronic acid, 1-tert-butoxycarbonyl-5-tert-butyl dimethyl silyl oxy-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyl oxy indol-2-ylboronic acid, 1-boc-5-tbdmso-2-indoleboronic acid, 1-boc-5-tert-butyldimethylsiloxy indole-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy indole, 5-tert-butyldimethylsilyloxy-1h-indole-2-boronic acid,n-boc protected 98
B(C1=CC2=C(N1C(=O)OC(C)(C)C)C=CC(=C2)O[Si](C)(C)C(C)(C)C)(O)O
391.346
391.35
335649-61-1
C19H30BNO5Si
250 mg
NXKIUYOUKLIWLP-UHFFFAOYSA-N
[5-[tert-butyl(dimethyl)silyl]oxy-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
22285134
98%

Chemical Identifiers

335649-61-1
391.346
NXKIUYOUKLIWLP-UHFFFAOYSA-N
22285134
B(C1=CC2=C(N1C(=O)OC(C)(C)C)C=CC(=C2)O[Si](C)(C)C(C)(C)C)(O)O
C19H30BNO5Si
MFCD08689532
5-tert-butyldimethylsilyloxy-1h-indole-2-boronic acid, n-boc protected, 1-boc-5-tbdmso-indole-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy-1h-indol-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy-1h-indol-2-ylboronic acid, 1-tert-butoxycarbonyl-5-tert-butyl dimethyl silyl oxy-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyl oxy indol-2-ylboronic acid, 1-boc-5-tbdmso-2-indoleboronic acid, 1-boc-5-tert-butyldimethylsiloxy indole-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy indole, 5-tert-butyldimethylsilyloxy-1h-indole-2-boronic acid,n-boc protected 98
[5-[tert-butyl(dimethyl)silyl]oxy-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions