missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Boc-6-methoxyindole-2-boronic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H52482MD

 View more versions of this product

Catalog No. AAH52482MD


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Boc-6-methoxyindole-2-boronic acid
100°C to 103°C
MFCD06659830
1-boc-6-methoxyindole-2-boronic acid, 1-tert-butoxycarbonyl-6-methoxy-1h-indol-2-yl boronic acid, 6-methoxy-1h-indole-2-boronic acid, n-boc protected, 1-boc-6-methoxy-2-indoleboronic acid, 6-methoxyindole-2-boronic acid, boc protected, 1-tert-butoxycarbonyl-6-methoxyindol-2-ylboronic acid, 1-t-butoxycarbonyl-6-methoxyindole-2-boronic acid, 1h-indole-1-carboxylic acid, 2-borono-6-methoxy-, 1-1,1-dimethylethyl ester, 1h-indole-1-carboxylicacid, 2-borono-6-methoxy-, 1-1,1-dimethylethyl ester
B(C1=CC2=C(N1C(=O)OC(C)(C)C)C=C(C=C2)OC)(O)O
291.11
291.11
850568-65-9
C14H18BNO5
250 mg
ATIJYIUOXDCYTN-UHFFFAOYSA-N
[6-methoxy-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
22309457
98%

Chemical Identifiers

850568-65-9
291.11
ATIJYIUOXDCYTN-UHFFFAOYSA-N
22309457
B(C1=CC2=C(N1C(=O)OC(C)(C)C)C=C(C=C2)OC)(O)O
C14H18BNO5
MFCD06659830
1-boc-6-methoxyindole-2-boronic acid, 1-tert-butoxycarbonyl-6-methoxy-1h-indol-2-yl boronic acid, 6-methoxy-1h-indole-2-boronic acid, n-boc protected, 1-boc-6-methoxy-2-indoleboronic acid, 6-methoxyindole-2-boronic acid, boc protected, 1-tert-butoxycarbonyl-6-methoxyindol-2-ylboronic acid, 1-t-butoxycarbonyl-6-methoxyindole-2-boronic acid, 1h-indole-1-carboxylic acid, 2-borono-6-methoxy-, 1-1,1-dimethylethyl ester, 1h-indole-1-carboxylicacid, 2-borono-6-methoxy-, 1-1,1-dimethylethyl ester
[6-methoxy-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions