Learn More
1-Boc-imidazole, 98%, Thermo Scientific Chemicals
$52.05 - $134.05
Chemical Identifiers
CAS | 49761-82-2 |
---|---|
Molecular Formula | C8H12N2O2 |
Molecular Weight (g/mol) | 168.196 |
MDL Number | MFCD00014497 |
InChI Key | MTBKGWHHOBJMHJ-UHFFFAOYSA-N |
Synonym | n-boc-imidazole, tert-butyl 1h-imidazole-1-carboxylate, n-tert-butoxycarbonylimidazole, 1-tert-butoxycarbonyl imidazole, 1h-imidazole-1-carboxylic acid, 1,1-dimethylethyl ester, imidazole-1-carboxylic acid tert-butyl ester, tert-butyl imidazolecarboxylate, 1-boc-imidazole, acmc-209ki1, ksc237k5p |
PubChem CID | 521262 |
IUPAC Name | tert-butyl imidazole-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1C=CN=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0050606
|
Thermo Scientific Chemicals
L0050606 |
5 g |
Each for $52.05
|
|
|||||
AAL0050614
|
Thermo Scientific Chemicals
L0050614 |
25 g |
Each for $134.05
|
|
|||||
Description
1-Boc-imidazole is used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N?-carbonyldiimidazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Boc-imidazole is used in the preparation of water soluble polyrotaxane, via modification of α-cyclodextrin in the presence of KOH and N,N′-carbonyldiimidazole.
Solubility
Soluble in hexane, acetone, and DMF.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
49761-82-2 | |
168.196 | |
MTBKGWHHOBJMHJ-UHFFFAOYSA-N | |
521262 | |
CC(C)(C)OC(=O)N1C=CN=C1 |
C8H12N2O2 | |
MFCD00014497 | |
n-boc-imidazole, tert-butyl 1h-imidazole-1-carboxylate, n-tert-butoxycarbonylimidazole, 1-tert-butoxycarbonyl imidazole, 1h-imidazole-1-carboxylic acid, 1,1-dimethylethyl ester, imidazole-1-carboxylic acid tert-butyl ester, tert-butyl imidazolecarboxylate, 1-boc-imidazole, acmc-209ki1, ksc237k5p | |
tert-butyl imidazole-1-carboxylate |
Specifications
49761-82-2 | |
C8H12N2O2 | |
5 g | |
n-boc-imidazole, tert-butyl 1h-imidazole-1-carboxylate, n-tert-butoxycarbonylimidazole, 1-tert-butoxycarbonyl imidazole, 1h-imidazole-1-carboxylic acid, 1,1-dimethylethyl ester, imidazole-1-carboxylic acid tert-butyl ester, tert-butyl imidazolecarboxylate, 1-boc-imidazole, acmc-209ki1, ksc237k5p | |
MTBKGWHHOBJMHJ-UHFFFAOYSA-N | |
tert-butyl imidazole-1-carboxylate | |
521262 | |
98% |
44°C to 48°C | |
MFCD00014497 | |
607792 | |
Soluble in hexane,acetone,and DMF. | |
CC(C)(C)OC(=O)N1C=CN=C1 | |
168.196 | |
168.2 | |
1-Boc-imidazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 256-475-1
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only