missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Boc-L-nipecotic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5257514

 View more versions of this product

Catalog No. AAH5257514

Explore available promotions

Description

Description

1-Boc-L-nipecotic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-L-nipecotic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

1-Boc-L-nipecotic acid
165°C to 169°C
MFCD02179172
s-1-boc-piperidine-3-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, s-n-boc-piperidine-3-carboxylic acid, l-1-boc-nipecotic acid, s-boc-nipecotic acid, boc-s-nipecotic acid, boc-s-nip-oh, s-boc-nip, s-n-boc-nipecotic acid, boc-nip-oh
+57° (c=1 in Chloroform)
(3S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
6951168
97%
88495-54-9
C11H19NO4
25 g
NXILIHONWRXHFA-QMMMGPOBSA-N
CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O
229.276
229.28

Chemical Identifiers

88495-54-9
229.276
NXILIHONWRXHFA-QMMMGPOBSA-N
6951168
CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O
C11H19NO4
MFCD02179172
s-1-boc-piperidine-3-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, s-n-boc-piperidine-3-carboxylic acid, l-1-boc-nipecotic acid, s-boc-nipecotic acid, boc-s-nipecotic acid, boc-s-nip-oh, s-boc-nip, s-n-boc-nipecotic acid, boc-nip-oh
(3S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only