missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1-BOC-Piperidin-4-yl)acetic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 157688-46-5
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.31
InChI Key ZXFLMSIMHISJFV-UHFFFAOYSA-N
Synonym 1-boc-4-piperidylacetic acid, 1-boc-piperidin-4-ylacetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl acetic acid, n-boc-4-piperidineacetic acid, 1-boc-4-piperidineacetic acid, 2-1-tert-butoxy carbonyl piperidin-4-yl acetic acid, 1-tert-butoxycarbonyl piperidin-4-yl acetic acid, boc-cmp, boc-cmp-oh
PubChem CID 1512539
IUPAC Name 2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]acetic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC437500010
SDS View Documents
Thermo Scientific Chemicals
437500010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC437500050
SDS View Documents
Thermo Scientific Chemicals
437500050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

157688-46-5
243.31
1-boc-4-piperidylacetic acid, 1-boc-piperidin-4-ylacetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl acetic acid, n-boc-4-piperidineacetic acid, 1-boc-4-piperidineacetic acid, 2-1-tert-butoxy carbonyl piperidin-4-yl acetic acid, 1-tert-butoxycarbonyl piperidin-4-yl acetic acid, boc-cmp, boc-cmp-oh
2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]acetic acid
C12H21NO4
ZXFLMSIMHISJFV-UHFFFAOYSA-N
1512539
CC(C)(C)OC(=O)N1CCC(CC1)CC(=O)O

Specifications

157688-46-5
100.0
97%
C12H21NO4
1-boc-4-piperidylacetic acid, 1-boc-piperidin-4-ylacetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl acetic acid, n-boc-4-piperidineacetic acid, 1-boc-4-piperidineacetic acid, 2-1-tert-butoxy carbonyl piperidin-4-yl acetic acid, 1-tert-butoxycarbonyl piperidin-4-yl acetic acid, boc-cmp, boc-cmp-oh
CC(C)(C)OC(=O)N1CCC(CC1)CC(=O)O
243.31
243.31
(1-BOC-Piperidin-4-yl)acetic acid
96.0
Authentic
Glass bottle
1g
ZXFLMSIMHISJFV-UHFFFAOYSA-N
2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]acetic acid
1512539
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only