Learn More
1-Bromo-2,4-dichlorobenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1229814
Description
1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4'-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4′-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
1-Bromo-2,4-dichlorobenzene | |
26°C to 27°C | |
C6H3BrCl2 | |
25 g | |
2,4-dichlorobromobenzene, bromo 2--4-dichlorobenzene, 1-bromo-2,4-dichloro-benzene, benzene, 1-bromo-2,4-dichloro, pubchem3608, 2,4-dichlorobromobenze, 2,4-dichloro-bromobenzene, acmc-209a2k, 2,4-dichloro-1-bromobenzene, ksc490m9l | |
ISHYFWKKWKXXPL-UHFFFAOYSA-N | |
1-bromo-2,4-dichlorobenzene | |
70947 | |
98% |
1193-72-2 | |
235°C | |
MFCD00018333 | |
1635733 | |
Slightly soluble in water. | |
C1=CC(=C(C=C1Cl)Cl)Br | |
225.894 | |
225.91 |
Chemical Identifiers
1193-72-2 | |
225.894 | |
ISHYFWKKWKXXPL-UHFFFAOYSA-N | |
70947 | |
C1=CC(=C(C=C1Cl)Cl)Br |
C6H3BrCl2 | |
MFCD00018333 | |
2,4-dichlorobromobenzene, bromo 2--4-dichlorobenzene, 1-bromo-2,4-dichloro-benzene, benzene, 1-bromo-2,4-dichloro, pubchem3608, 2,4-dichlorobromobenze, 2,4-dichloro-bromobenzene, acmc-209a2k, 2,4-dichloro-1-bromobenzene, ksc490m9l | |
1-bromo-2,4-dichlorobenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-778-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only