missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromo-2,5-bis(trifluoromethyl)benzene 87.0+%, TCI America™
Supplier: TCI America B25455G
Specifications
1-Bromo-2,5-bis(trifluoromethyl)benzene | |
5°C | |
C8H3BrF6 | |
5 g | |
GFQNSGHVOFVTLC-UHFFFAOYSA-N | |
2-bromo-1,4-bis(trifluoromethyl)benzene | |
2734763 | |
≥87.0% (GC) |
7617-93-8 | |
158°C | |
MFCD03094336 | |
2,5-bis trifluoromethyl bromobenzene, 2-bromo-1,4-bis trifluoromethyl benzene, 1-bromo-2,5-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-bromobenzene, benzene, 2-bromo-1,4-bis trifluoromethyl, pubchem4264, acmc-1bisb, ksc495k7j, 2,5-bis-trifluoromethylbromobenzene | |
C1=CC(=C(C=C1C(F)(F)F)Br)C(F)(F)F | |
293.006 | |
293.01 | |
Liquid |
Chemical Identifiers
7617-93-8 | |
293.006 | |
GFQNSGHVOFVTLC-UHFFFAOYSA-N | |
2734763 | |
C1=CC(=C(C=C1C(F)(F)F)Br)C(F)(F)F |
C8H3BrF6 | |
MFCD03094336 | |
2,5-bis trifluoromethyl bromobenzene, 2-bromo-1,4-bis trifluoromethyl benzene, 1-bromo-2,5-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-bromobenzene, benzene, 2-bromo-1,4-bis trifluoromethyl, pubchem4264, acmc-1bisb, ksc495k7j, 2,5-bis-trifluoromethylbromobenzene | |
2-bromo-1,4-bis(trifluoromethyl)benzene |
Safety and Handling
TSCA : No