missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-butyne, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 387020050

 View more versions of this product

Catalog No. AC387020050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromo-2-butyne
97.5
Green-Yellow
40.0°C to 41.0°C
Authentic
Glass bottle
1.5070 to 1.5090 (20°C, 589nm)
MFCD00190233
1.473
LNNXOEHOXSYWLD-UHFFFAOYSA-N
1-bromobut-2-yne
2756862
98%
3355-28-0
100.0
1.4730g/mL
36°C
97.5% min. (GC)
C4H5Br
CH3CCCH2Br
5 g
1-bromo-2-butyne, 2-butyne, 1-bromo, 2-butynyl bromide, 1-bromo-but-2-yne, bromo-2-butyne, 1-bromo2-butyne, but-2-ynylbromide, l-bromo-2-butine, 1-bromo-2-butine, 4-bromo-2-butyne
CC#CCBr
132.99
132.99
Liquid

Chemical Identifiers

3355-28-0
132.99
LNNXOEHOXSYWLD-UHFFFAOYSA-N
2756862
CC#CCBr
C4H5Br
MFCD00190233
1-bromo-2-butyne, 2-butyne, 1-bromo, 2-butynyl bromide, 1-bromo-but-2-yne, bromo-2-butyne, 1-bromo2-butyne, but-2-ynylbromide, l-bromo-2-butine, 1-bromo-2-butine, 4-bromo-2-butyne
1-bromobut-2-yne
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only