missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-chloro-4-fluorobenzene, 98%, Thermo Scientific Chemicals

$76.70 - $76.70

Chemical Identifiers

CAS 110407-59-5
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD00142587
InChI Key LEFQPBAWVJEIJS-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobromobenzene, 4-bromo-3-chloro-1-fluorobenzene, 1-bromo-2-chloro-4-fluoro-benzene, 4-bromo-3-chlorofluorobenzene, 2-bromo-5-fluorochlorobenzene, benzene, 1-bromo-2-chloro-4-fluoro, pubchem3755, acmc-1bnv0, 2-chloro-4-fluorobromobezene, ksc494m7f
PubChem CID 2733406
IUPAC Name 1-bromo-2-chloro-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Cl)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2553806
View Documents
Thermo Scientific Chemicals
B2553806
5 g
Each for $76.70
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

110407-59-5
209.442
LEFQPBAWVJEIJS-UHFFFAOYSA-N
2733406
C1=CC(=C(C=C1F)Cl)Br
C6H3BrClF
MFCD00142587
2-chloro-4-fluorobromobenzene, 4-bromo-3-chloro-1-fluorobenzene, 1-bromo-2-chloro-4-fluoro-benzene, 4-bromo-3-chlorofluorobenzene, 2-bromo-5-fluorochlorobenzene, benzene, 1-bromo-2-chloro-4-fluoro, pubchem3755, acmc-1bnv0, 2-chloro-4-fluorobromobezene, ksc494m7f
1-bromo-2-chloro-4-fluorobenzene

Specifications

110407-59-5
1.552
5 g
LEFQPBAWVJEIJS-UHFFFAOYSA-N
1-bromo-2-chloro-4-fluorobenzene
2733406
98%
C6H3BrClF
MFCD00142587
2-chloro-4-fluorobromobenzene, 4-bromo-3-chloro-1-fluorobenzene, 1-bromo-2-chloro-4-fluoro-benzene, 4-bromo-3-chlorofluorobenzene, 2-bromo-5-fluorochlorobenzene, benzene, 1-bromo-2-chloro-4-fluoro, pubchem3755, acmc-1bnv0, 2-chloro-4-fluorobromobezene, ksc494m7f
C1=CC(=C(C=C1F)Cl)Br
209.442
209.45
1-Bromo-2-chloro-4-fluorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only