missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloro-4-fluorobenzene 98.0+%, TCI America™

Supplier:  TCI America B334925G

 View more versions of this product

Catalog No. B334925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Bromo-2-chloro-4-fluorobenzene
Yellow
MFCD00142587
2-chloro-4-fluorobromobenzene, 4-bromo-3-chloro-1-fluorobenzene, 1-bromo-2-chloro-4-fluoro-benzene, 4-bromo-3-chlorofluorobenzene, 2-bromo-5-fluorochlorobenzene, benzene, 1-bromo-2-chloro-4-fluoro, pubchem3755, acmc-1bnv0, 2-chloro-4-fluorobromobezene, ksc494m7f
C1=CC(=C(C=C1F)Cl)Br
209.442
209.44
Liquid
110407-59-5
C6H3BrClF
25 g
LEFQPBAWVJEIJS-UHFFFAOYSA-N
1-bromo-2-chloro-4-fluorobenzene
2733406
≥98.0% (GC)

Chemical Identifiers

110407-59-5
209.442
LEFQPBAWVJEIJS-UHFFFAOYSA-N
2733406
C1=CC(=C(C=C1F)Cl)Br
C6H3BrClF
MFCD00142587
2-chloro-4-fluorobromobenzene, 4-bromo-3-chloro-1-fluorobenzene, 1-bromo-2-chloro-4-fluoro-benzene, 4-bromo-3-chlorofluorobenzene, 2-bromo-5-fluorochlorobenzene, benzene, 1-bromo-2-chloro-4-fluoro, pubchem3755, acmc-1bnv0, 2-chloro-4-fluorobromobezene, ksc494m7f
1-bromo-2-chloro-4-fluorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions