Learn More
1-Bromo-2-chlorobenzene, 98+%, Thermo Scientific Chemicals
$100.90 - $1269.36
Chemical Identifiers
CAS | 694-80-4 |
---|---|
Molecular Formula | C6H4BrCl |
Molecular Weight (g/mol) | 191.452 |
MDL Number | MFCD00000532 |
InChI Key | QBELEDRHMPMKHP-UHFFFAOYSA-N |
Synonym | 2-bromochlorobenzene, o-bromochlorobenzene, 2-chlorobromobenzene, o-chlorobromobenzene, benzene, 1-bromo-2-chloro, 2-bromo-1-chlorobenzene, 1-chloro-2-bromobenzene, benzene, bromochloro, unii-pec7z3yx6p, 1-bromo-2-chloro-benzene |
PubChem CID | 12754 |
IUPAC Name | 1-bromo-2-chlorobenzene |
SMILES | C1=CC=C(C(=C1)Cl)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1178218
|
Thermo Scientific Chemicals
A1178218 |
50 g |
Each for $100.90
|
|
|||||
AAA1178230
|
Thermo Scientific Chemicals
A1178230 |
250 g |
Each for $324.92
|
|
|||||
AAA117820B
|
Thermo Scientific Chemicals
A117820B |
1000 g |
Each for $1,269.36
|
|
|||||
Description
1-Bromo-2-chlorobenzene finds application in Gas Chromatography and Liquid Chromatography analysis. It is also used as an intermediate in pharmaceutical and synthetic chemistry. Further, it acts as a reagent in the preparation of arylpiperidines and aryltetrahydropyridines as 5-HT2C agonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-2-chlorobenzene finds application in Gas Chromatography and Liquid Chromatography analysis. It is also used as an intermediate in pharmaceutical and synthetic chemistry. Further, it acts as a reagent in the preparation of arylpiperidines and aryltetrahydropyridines as 5-HT2C agonists.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
694-80-4 | |
191.452 | |
QBELEDRHMPMKHP-UHFFFAOYSA-N | |
12754 | |
C1=CC=C(C(=C1)Cl)Br |
C6H4BrCl | |
MFCD00000532 | |
2-bromochlorobenzene, o-bromochlorobenzene, 2-chlorobromobenzene, o-chlorobromobenzene, benzene, 1-bromo-2-chloro, 2-bromo-1-chlorobenzene, 1-chloro-2-bromobenzene, benzene, bromochloro, unii-pec7z3yx6p, 1-bromo-2-chloro-benzene | |
1-bromo-2-chlorobenzene |
Specifications
-13°C | |
1.649 | |
50 g | |
C6H4BrCl | |
1855303 | |
Immiscible with water. | |
C1=CC=C(C(=C1)Cl)Br | |
191.452 | |
191.46 | |
1-Bromo-2-chlorobenzene |
694-80-4 | |
203°C to 205°C | |
1.5815 | |
MFCD00000532 | |
2-bromochlorobenzene, o-bromochlorobenzene, 2-chlorobromobenzene, o-chlorobromobenzene, benzene, 1-bromo-2-chloro, 2-bromo-1-chlorobenzene, 1-chloro-2-bromobenzene, benzene, bromochloro, unii-pec7z3yx6p, 1-bromo-2-chloro-benzene | |
QBELEDRHMPMKHP-UHFFFAOYSA-N | |
1-bromo-2-chlorobenzene | |
12754 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H332-H315-H319-H335
EINECSNumber : 211-775-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only