missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-fluorobenzene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1063514

 View more versions of this product

Catalog No. AAA1063514


Explore available promotions

Description

Description

1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents .
Specifications

Specifications

1-Bromo-2-fluorobenzene
-8°C
155°C to 157°C
Aromatic
1.534
25 g
1071819
Insoluble in water.
C1=CC=C(C(=C1)F)Br
175
175.01
1072-85-1
1.601
43°C (109°F)
C6H4BrF
MFCD00000282
UN1993
2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
IPWBFGUBXWMIPR-UHFFFAOYSA-N
1-bromo-2-fluorobenzene
61259
99%

Chemical Identifiers

1072-85-1
175
IPWBFGUBXWMIPR-UHFFFAOYSA-N
61259
C1=CC=C(C(=C1)F)Br
C6H4BrF
MFCD00000282
2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
1-bromo-2-fluorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H319
Flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 214-018-3

RTECSNumber : DA1050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only