Learn More
1-Bromo-2-fluorobenzene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1063536
Description
1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents .
Specifications
1-Bromo-2-fluorobenzene | |
-8°C | |
155°C to 157°C | |
Aromatic | |
1.534 | |
500 g | |
1071819 | |
Insoluble in water. | |
C1=CC=C(C(=C1)F)Br | |
175 | |
175.01 |
1072-85-1 | |
1.601 | |
43°C (109°F) | |
C6H4BrF | |
MFCD00000282 | |
UN1993 | |
2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro | |
IPWBFGUBXWMIPR-UHFFFAOYSA-N | |
1-bromo-2-fluorobenzene | |
61259 | |
99% |
Chemical Identifiers
1072-85-1 | |
175 | |
IPWBFGUBXWMIPR-UHFFFAOYSA-N | |
61259 | |
C1=CC=C(C(=C1)F)Br |
C6H4BrF | |
MFCD00000282 | |
2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro | |
1-bromo-2-fluorobenzene |
Safety and Handling
GHS H Statement
H226-H302-H319
Flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 214-018-3
RTECSNumber : DA1050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only