missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-iodobenzene, 99%, stabilized, Thermo Scientific Chemicals

$85.31 - $248.89

Chemical Identifiers

CAS 583-55-1
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.9
MDL Number MFCD00001030
InChI Key OIRHKGBNGGSCGS-UHFFFAOYSA-N
Synonym 2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
PubChem CID 11415
IUPAC Name 1-bromo-2-iodobenzene
SMILES C1=CC=C(C(=C1)Br)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC221950050
SDS View Documents Promotion Details
Thermo Scientific Chemicals
221950050
5 g Glass bottle
Each for $85.31
 
AC221950250
SDS View Documents Promotion Details
Thermo Scientific Chemicals
221950250
25 g Glass bottle
Each for $248.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

583-55-1
282.9
OIRHKGBNGGSCGS-UHFFFAOYSA-N
11415
C1=CC=C(C(=C1)Br)I
C6H4BrI
MFCD00001030
2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
1-bromo-2-iodobenzene

Specifications

583-55-1
100.0
Yellow
120°C to 121°C (15.0 mmHg)
Authentic
Glass bottle
1.6620 to 1.664
MFCD00001030
05, 223
2.203
OIRHKGBNGGSCGS-UHFFFAOYSA-N
1-bromo-2-iodobenzene
copper
282.9
Liquid
98.5
9°C to 10°C
2.2030g/mL
>110°C
98.5% min. (GC)
C6H4BrI
BrC6H4I
5 g
01,77
2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
C1=CC=C(C(=C1)Br)I
282.9
11415
99%
1-Bromo-2-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-508-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only